Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V1E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039366.1 | NP_001034455.1 | 4 | 441 | ||
NM_001696.3 | NP_001687.1 | 5 | 507 | |||
GenBank mRNA | AK294623.1 | - | 4 | 368 | ||
AK312394.1 | - | 5 | 428 | |||
BC004443.1 | - | 5 | 409 | 2820930 | ||
BI546032.1 | - | 5 | 459 | |||
BQ888820.1 | - | 4 | 386 | |||
BT007128.1 | - | 5 | 320 | |||
CK005516.1 | - | 5 | 510 | |||
CR456385.1 | - | 5 | 373 | |||
X71491.1 | - | 5 | 376 | |||
X76228.1 | - | 5 | 395 |