Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001710.5 | NP_001701.2 | 2, 3 | 571 | ||
GenBank mRNA | AF349679.1 | - | 2, 3 | 293 | ||
AK130533.1 | - | 2, 3 | 500 | |||
AK223400.1 | - | 2, 3 | 419 | |||
BC004143.2 | - | 2, 3 | 428 | 2959705 | ||
BC007990.2 | - | 2, 3 | 428 | 2959706 | ||
CD689714.1 | - | 2, 3 | 438 | |||
K01566.1 | - | 1, 2 | 32 | |||
L15702.1 | - | 2, 3 | 333 | |||
S67310.1 | - | 2, 3 | 404 | |||
X00284.1 | - | 2, 3 | 248 | |||
X72875.1 | - | 2, 3 | 293 |