Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk1b11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010640.1 | NP_034770.1 | 345 | |||
GenBank mRNA | BC013660.1 | - | 345 | 4167568 |