Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phgdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016966.3 | NP_058662.2 | 1 | 241 | ||
GenBank mRNA | AK076815.1 | - | 1 | 197 | ||
AK147946.1 | - | 1 | 189 | |||
AK169684.1 | - | 1 | 197 | |||
BC027566.1 | - | 116 | ||||
BC086668.1 | - | 1 | 149 | 30633243 | ||
BC110673.1 | - | 1 | 183 | 6311873 | ||
BY290664.1 | - | 1 | 241 |