Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAST SIRNA Assays ›
Gene Symbol:
CAST
Gene Name:
calpastatin
Gene Aliases:
BS-17, PLACK
Chromosome Location:
Chr.5: 96662037 - 96774683 on Build GRCh38
UniGene ID:
Hs.436186
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
10294
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAST
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042440.3
NP_001035905.1
15
1390
NM_001190442.1
NP_001177371.1
16
1262
NM_001284212.2
NP_001271141.1
13
1041
NM_001284213.2
NP_001271142.1
12
948
NM_173060.3
NP_775083.1
14
1098
NR_104285.1
2
254
XM_006714696.3
XP_006714759.1
18
1404
XM_006714697.3
XP_006714760.1
17
1421
XM_006714698.3
XP_006714761.1
16
1382
XM_006714699.3
XP_006714762.1
17
1345
XM_006714700.3
XP_006714763.1
16
1364
XM_006714701.3
XP_006714764.1
16
1380
XM_006714702.3
XP_006714765.1
16
1256
XM_006714703.3
XP_006714766.1
15
1341
XM_006714704.3
XP_006714767.1
16
1289
XM_006714705.3
XP_006714768.1
15
1298
XM_006714706.3
XP_006714769.1
15
1216
XM_006714707.3
XP_006714770.1
15
1167
XM_006714708.3
XP_006714771.1
14
1127
XM_006714709.3
XP_006714772.1
14
1090
XM_006714710.3
XP_006714773.1
13
1045
XM_006714711.3
XP_006714774.1
13
1090
XM_006714712.3
XP_006714775.1
12
1037
XM_006714713.3
XP_006714776.1
2
254
XM_006714714.3
XP_006714777.1
2
254
XM_006714715.3
XP_006714778.1
3
980
XM_011543654.2
XP_011541956.1
16
1340
XM_011543656.2
XP_011541958.1
15
1284
XM_011543657.2
XP_011541959.1
15
1166
XM_011543658.2
XP_011541960.1
13
1019
XM_017009911.1
XP_016865400.1
16
1382
XM_017009912.1
XP_016865401.1
17
1393
XM_017009913.1
XP_016865402.1
17
1362
XM_017009914.1
XP_016865403.1
16
1364
XM_017009915.1
XP_016865404.1
17
1366
XM_017009916.1
XP_016865405.1
16
1380
XM_017009917.1
XP_016865406.1
16
1298
XM_017009918.1
XP_016865407.1
15
1325
XM_017009920.1
XP_016865409.1
15
1208
XM_017009921.1
XP_016865410.1
15
1225
XM_017009922.1
XP_016865411.1
14
1259
XM_017009923.1
XP_016865412.1
14
1259
XM_017009925.1
XP_016865414.1
14
1177
XM_017009926.1
XP_016865415.1
14
1177
XM_017009927.1
XP_016865416.1
15
1166
XM_017009928.1
XP_016865417.1
14
1165
XM_017009929.1
XP_016865418.1
14
1084
XM_017009930.1
XP_016865419.1
13
1051
XM_017009931.1
XP_016865420.1
13
1045
XM_017009932.1
XP_016865421.1
12
1108
GenBank mRNA
AB208816.1
-
16
1226
AF327443.1
-
11
940
AK292289.1
-
2
254
AK292356.1
-
2
227
AK296919.1
-
16
1220
AK297879.1
-
14
1040
AK298541.1
-
15
1167
AK299384.1
-
15
1071
AK299402.1
-
15
1147
AK300624.1
-
12
931
AK302206.1
-
3
254
AK303836.1
-
8
621
AK303900.1
-
11
834
AK311585.1
-
3
432
AK311598.1
-
3
308
AK314290.1
-
13
1043
AK315146.1
-
13
1042
AK315673.1
-
2
202
AL832174.1
-
14
1083
AL832349.1
-
14
1086
AY189741.1
-
4
253
BC013579.1
-
15
1093
3878564
BC078149.1
-
10
676
BT009783.1
-
12
875
BX648397.1
-
13
992
D16217.1
-
15
1160
DB033043.1
-
2
202
U26724.2
-
6
523
U31345.1
-
4
270
U58996.2
-
2
233
Back To Top
More Information
Panther Classification:
Molecular Function -
protein-binding activity modulator
protease inhibitor
Gene Ontology Categories:
Function(s)
Process(es)
liver development
egg activation
brain development
myoblast fusion
aging
negative regulation of endopeptidase activity
organ regeneration
myoblast differentiation
negative regulation of cell cycle arrest
cell-cell adhesion
negative regulation of type B pancreatic cell apoptotic process
protease binding
endopeptidase inhibitor activity
protein binding
calcium-dependent cysteine-type endopeptidase inhibitor activity
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0