Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122960.1 | NP_001116432.1 | 2 | 222 | ||
NM_026070.3 | NP_080346.1 | 2 | 222 | |||
NM_027487.1 | NP_081763.1 | 2 | 222 | |||
GenBank mRNA | AK004206.1 | - | 2 | 137 | ||
AK009358.1 | - | 2 | 118 | |||
AK013835.1 | - | 2 | 227 | |||
AK017732.1 | - | 2 | 150 | |||
AK028599.1 | - | 1 | 655 | |||
AK158822.1 | - | 2 | 223 | |||
BC029256.1 | - | 2 | 119 | 3661472 | ||
CK795584.1 | - | 2 | 174 |