Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2afb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026627.1 | NP_080903.1 | 1 | 181 | ||
GenBank mRNA | AK006586.1 | - | 1 | 167 | ||
AK015538.1 | - | 1 | 172 | |||
AK018839.1 | - | 1 | 181 | |||
AK018883.1 | - | 1 | 103 | |||
AK018922.1 | - | 1 | 167 | |||
AK018951.1 | - | 1 | 167 | |||
AK018962.1 | - | 1 | 167 | |||
AK160415.1 | - | 1 | 167 | |||
BC048736.1 | - | 1 | 145 | |||
BC061062.1 | - | 1 | 193 | 6774311 | ||
EU079024.1 | - | 1 | 108 |