Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arxes1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029541.3 | NP_083817.1 | 1 | 298 | ||
GenBank mRNA | AK018307.1 | - | 1 | 298 | ||
BC016270.1 | - | 1 | 277 | |||
BY287880.1 | - | 1 | 299 |
Gene Symbol: Arxes2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029823.2 | NP_084099.1 | 1 | 298 | ||
GenBank mRNA | AK013740.1 | - | 1 | 295 | ||
AK077432.1 | - | 1 | 272 | |||
BC030351.1 | - | 1 | 298 | |||
BY773138.1 | - | 1 | 299 |