Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029576.3 | NP_083852.1 | 3 | 169 | ||
GenBank mRNA | AB232584.1 | - | 2 | 111 | ||
AK003609.1 | - | 3 | 170 | |||
AK042020.1 | - | 3 | 171 | |||
AK156726.1 | - | 3 | 192 | |||
AK208578.1 | - | 1 | 33 | |||
AK215506.1 | - | 192 | ||||
BC016408.1 | - | 3 | 164 | 4242206 |