Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2c50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167875.1 | NP_001161347.1 | 1 | 161 | ||
NM_001167877.1 | NP_001161349.1 | 1 | 161 | |||
NM_134144.2 | NP_598905.2 | 1 | 161 | |||
GenBank mRNA | AK149466.1 | - | 1 | 106 | ||
AY206873.1 | - | 1 | 140 | |||
BC011222.1 | - | 1 | 144 | 4195223 | ||
BC051050.1 | - | 1 | 152 | 5052536 | ||
BY086770.1 | - | 1 | 164 |