Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145215.2 | NP_660250.1 | 1 | 522 | ||
GenBank mRNA | AF412033.1 | - | 1 | 522 | ||
AK004244.1 | - | 1 | 57 | |||
BC056624.1 | - | 1 | 365 | |||
BC069865.1 | - | 1 | 115 | |||
BC069866.1 | - | 1 | 144 | 6478980 |