Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148953.2 | NP_683755.1 | 1 | 221 | ||
GenBank mRNA | AF403044.1 | - | 1 | 221 | ||
BC127245.1 | - | 1 | 217 | |||
BC127246.1 | - | 1 | 220 | |||
BC152348.1 | - | 1 | 221 | |||
BC152349.1 | - | 1 | 221 | 40104909 |