Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174875.3 | NP_777364.3 | 129 | |||
GenBank mRNA | AK041379.1 | - | 128 | |||
AK162404.1 | - | 131 | ||||
BB638045.1 | - | 128 | ||||
BC089500.1 | - | 130 | 30286094 | |||
BY287162.1 | - | 130 | ||||
CK023195.1 | - | 172 |