Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001647.3 | NP_001638.1 | 3 | 559 | ||
GenBank mRNA | AK294523.1 | - | 3 | 375 | ||
AK312090.1 | - | 3 | 294 | |||
BC007402.2 | - | 3 | 274 | 3050012 | ||
BT019860.1 | - | 2 | 197 | |||
BT019861.1 | - | 2 | 197 | |||
BU195270.1 | - | 3 | 472 | |||
CR456838.1 | - | 2 | 197 | |||
CR541773.1 | - | 2 | 197 | |||
DA892855.1 | - | 3 | 559 | |||
J02611.1 | - | 3 | 258 |