Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002751.6 | NP_002742.3 | 10 | 913 | ||
NR_110887.1 | 10 | 913 | ||||
XM_011530706.1 | XP_011529008.1 | 8 | 637 | |||
GenBank mRNA | AF001008.1 | - | 10 | 812 | ||
AF001174.1 | - | 10 | 818 | |||
AF031135.1 | - | 10 | 827 | |||
AK291845.1 | - | 10 | 864 | |||
BC027933.1 | - | 10 | 870 | 5199628 | ||
CR456514.1 | - | 10 | 814 | |||
U53442.1 | - | 10 | 855 | |||
U92268.1 | - | 10 | 816 | |||
Y14440.1 | - | 10 | 818 |