Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193511.1 | NP_001180440.1 | 5 | 1155 | ||
NM_006301.3 | NP_006292.3 | 6 | 1056 | |||
XM_005269138.3 | XP_005269195.1 | 5 | 1083 | |||
XM_005269140.2 | XP_005269197.1 | 5 | 996 | |||
XM_006719588.3 | XP_006719651.1 | 5 | 977 | |||
XM_011538725.2 | XP_011537027.1 | 5 | 1960 | |||
XM_017019956.1 | XP_016875445.1 | 6 | 984 | |||
GenBank mRNA | AB209453.1 | - | 1 | 1316 | ||
AK094195.1 | - | 5 | 951 | |||
AK314530.1 | - | 5 | 1303 | |||
BC050050.1 | - | 5 | 1156 | 4824512 | ||
U07358.1 | - | 6 | 883 |