Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014911.3 | NP_055726.3 | 3 | 704 | ||
GenBank mRNA | AB028971.1 | - | 3 | 583 | ||
AK056043.1 | - | 3 | 511 | |||
AK310951.1 | - | 3 | 507 | |||
BC002695.2 | - | 2 | 239 | |||
BC104842.1 | - | 2 | 223 | 8143845 | ||
BC111965.1 | - | 2 | 223 | 8327433 | ||
BC143710.1 | - | 2 | 223 | 9052224 | ||
DB285703.1 | - | 3 | 512 |