Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other STK33 SIRNA Assays ›
Gene Symbol:
STK33
Gene Name:
serine/threonine kinase 33
Gene Aliases:
-
Chromosome Location:
Chr.11: 8335029 - 8594289 on Build GRCh38
UniGene ID:
Hs.501833
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
103367
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: STK33
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289058.1
NP_001275987.1
4
483
NM_001289061.1
NP_001275990.1
3
748
NM_030906.3
NP_112168.1
4
897
XM_011520288.2
XP_011518590.1
4
1975
XM_011520290.2
XP_011518592.1
6
894
XM_011520292.2
XP_011518594.1
4
696
XM_011520293.2
XP_011518595.1
4
725
XM_011520294.1
XP_011518596.1
3
690
XM_011520295.2
XP_011518597.1
3
2786
XM_011520297.2
XP_011518599.1
3
463
XM_017018143.1
XP_016873632.1
6
954
XM_017018144.1
XP_016873633.1
6
856
XM_017018145.1
XP_016873634.1
5
2888
XM_017018146.1
XP_016873635.1
6
1018
XM_017018147.1
XP_016873636.1
5
2815
XM_017018148.1
XP_016873637.1
4
2747
XM_017018149.1
XP_016873638.1
5
899
XM_017018150.1
XP_016873639.1
5
754
XM_017018151.1
XP_016873640.1
5
713
XM_017018152.1
XP_016873641.1
3
1044
XM_017018153.1
XP_016873642.1
3
642
XM_017018154.1
XP_016873643.1
3
501
XM_017018155.1
XP_016873644.1
3
1056
XM_017018156.1
XP_016873645.1
3
502
XM_017018157.1
XP_016873646.1
3
267
XM_017018158.1
XP_016873647.1
4
553
XM_017018159.1
XP_016873648.1
5
2891
XM_017018160.1
XP_016873649.1
3
690
XM_017018161.1
XP_016873650.1
5
2897
XM_017018162.1
XP_016873651.1
4
500
XM_017018163.1
XP_016873652.1
3
491
XM_017018164.1
XP_016873653.1
3
377
XM_017018165.1
XP_016873654.1
4
456
GenBank mRNA
AJ303380.2
-
2
1241
AK093251.1
-
3
606
AK293193.1
-
4
349
BC031231.1
-
4
845
5296502
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein autophosphorylation
protein serine/threonine kinase activity
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0