Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002738.6 | NP_002729.2 | 8 | 1048 | ||
NM_212535.2 | NP_997700.1 | 8 | 1048 | |||
GenBank mRNA | BC036472.1 | - | 8 | 1008 | 5260547 | |
M13975.1 | - | 8 | 915 | |||
X06318.1 | - | 8 | 987 | |||
X07109.1 | - | 8 | 987 |