Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002742.2 | NP_002733.2 | 3, 4 | 724 | ||
XM_005267859.1 | XP_005267916.1 | 3, 4 | 658 | |||
XM_011536964.2 | XP_011535266.1 | 3, 4 | 3080 | |||
XM_011536965.2 | XP_011535267.1 | 3, 4 | 5130 | |||
XM_017021461.1 | XP_016876950.1 | 3, 4 | 435 | |||
XM_017021462.1 | XP_016876951.1 | 4, 5 | 421 | |||
GenBank mRNA | AK314170.1 | - | 3, 4 | 660 | ||
CN412379.1 | - | 3, 4 | 312 | |||
X75756.1 | - | 3, 4 | 778 |