Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278273.1 | NP_001265202.1 | 7, 8 | 655 | ||
NM_182691.2 | NP_872633.1 | 5, 6 | 708 | |||
NM_182692.2 | NP_872634.1 | 6, 7 | 631 | |||
XM_005250549.3 | XP_005250606.2 | 7, 8 | 890 | |||
XM_005250550.3 | XP_005250607.1 | 6, 7 | 635 | |||
XM_005250551.4 | XP_005250608.1 | 7, 8 | 683 | |||
XM_006716098.2 | XP_006716161.1 | 7, 8 | 650 | |||
XM_011516536.2 | XP_011514838.1 | 5, 6 | 646 | |||
XM_011516538.2 | XP_011514840.1 | 6, 7 | 519 | |||
XM_017012560.1 | XP_016868049.1 | 7, 8 | 705 | |||
XM_017012561.1 | XP_016868050.1 | 6, 7 | 555 | |||
XM_017012562.1 | XP_016868051.1 | 7, 8 | 889 | |||
XM_017012563.1 | XP_016868052.1 | 7, 8 | 890 | |||
XM_017012564.1 | XP_016868053.1 | 5, 6 | 694 | |||
XM_017012565.1 | XP_016868054.1 | 7, 8 | 654 | |||
XM_017012566.1 | XP_016868055.1 | 7, 8 | 691 | |||
GenBank mRNA | AY354201.1 | - | 6, 7 | 696 | ||
BC035214.1 | - | 5, 6 | 627 | 4778251 | ||
BC068547.1 | - | 7, 8 | 655 | 5265599 | ||
BI918658.1 | - | 6, 7 | 628 | |||
BM805927.1 | - | 682 | ||||
U88666.1 | - | 5, 6 | 661 |