Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYRK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004023.1 | NP_001004023.1 | 4 | 835 | ||
NM_003582.2 | NP_003573.2 | 3 | 741 | |||
XM_005273315.4 | XP_005273372.1 | 4 | 679 | |||
XM_011510061.2 | XP_011508363.1 | 3 | 575 | |||
GenBank mRNA | AF186773.1 | - | 3 | 718 | ||
AF186774.1 | - | 4 | 984 | |||
AF327561.1 | - | 4 | 613 | |||
AK291189.1 | - | 4 | 835 | |||
AK308082.1 | - | 3 | 741 | |||
AK313245.1 | - | 3 | 741 | |||
AY590695.1 | - | 3 | 740 | |||
BC015501.1 | - | 4 | 792 | 3913541 | ||
Y12735.2 | - | 3 | 721 |