Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007371.3 | NP_031397.1 | 6 | 1155 | ||
XM_006717291.2 | XP_006717354.1 | 6 | 1287 | |||
XM_011519052.2 | XP_011517354.1 | 6 | 1506 | |||
XM_017015165.1 | XP_016870654.1 | 5 | 1080 | |||
XM_017015166.1 | XP_016870655.1 | 3 | 472 | |||
GenBank mRNA | AK299519.1 | - | 6 | 1190 | ||
AY513270.1 | - | 5 | 985 | |||
BC032124.2 | - | 6 | 1265 | 4856840 | ||
D26362.1 | - | 6 | 1106 | |||
KR633047.1 | - | 5 | 967 |