Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZAK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016653.2 | NP_057737.2 | 2 | 328 | ||
NM_133646.2 | NP_598407.1 | 2 | 328 | |||
XM_005246640.2 | XP_005246697.1 | 2 | 374 | |||
XM_017004323.1 | XP_016859812.1 | 2 | 374 | |||
XM_017004324.1 | XP_016859813.1 | 2 | 398 | |||
GenBank mRNA | AB030034.1 | - | 2 | 303 | ||
AB041883.1 | - | 2 | 398 | |||
AB049733.1 | - | 1 | 128 | |||
AB049734.1 | - | 1 | 128 | |||
AB208974.1 | - | 2 | 264 | |||
AF238255.1 | - | 1 | 128 | |||
AF251441.1 | - | 2 | 206 | |||
AF325454.1 | - | 1 | 133 | |||
AF465843.1 | - | 2 | 297 | |||
AF480461.1 | - | 2 | 323 | |||
AF480462.1 | - | 2 | 323 | |||
AK056310.1 | - | 2 | 329 | |||
AK291825.1 | - | 2 | 220 | |||
BC001401.2 | - | 2 | 258 | 3051317 | ||
DA993056.1 | - | 2 | 329 |