Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HGS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004712.4 | NP_004703.1 | 10, 11 | 974 | ||
XM_011525463.2 | XP_011523765.1 | 10, 11 | 923 | |||
XM_017025297.1 | XP_016880786.1 | 11, 12 | 1131 | |||
XM_017025298.1 | XP_016880787.1 | 9, 10 | 854 | |||
GenBank mRNA | AF260566.1 | - | 10, 11 | 843 | ||
AK303800.1 | - | 10, 11 | 890 | |||
BC003565.1 | - | 10, 11 | 916 | 3544197 | ||
BT009754.1 | - | 10, 11 | 839 | |||
D84064.1 | - | 10, 11 | 899 | |||
KU178466.1 | - | 10, 11 | 839 | |||
U43895.1 | - | 10, 11 | 914 |