Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260504.1 | NP_001247433.1 | 16 | 2443 | ||
NM_001260505.1 | NP_001247434.1 | 16 | 2167 | |||
NM_031414.4 | NP_113602.2 | 16 | 2167 | |||
NM_032944.3 | NP_116562.2 | 16 | 2411 | |||
NR_048542.1 | 16 | 2159 | ||||
XM_006715758.3 | XP_006715821.1 | 14 | 1547 | |||
XM_011515449.2 | XP_011513751.2 | 16 | 2093 | |||
XM_011515450.1 | XP_011513752.1 | 16 | 2414 | |||
XM_011515452.2 | XP_011513754.2 | 14 | 1560 | |||
GenBank mRNA | AF285599.1 | - | 16 | 2063 | ||
AF332194.1 | - | 16 | 2446 | |||
AK131214.1 | - | 7 | 869 | |||
AK292474.1 | - | 17 | 2059 | |||
AK302689.1 | - | 16 | 2059 | |||
BC033018.1 | - | 16 | 2061 | |||
BC059374.1 | - | 16 | 2074 | 5268051 |