Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138316.3 | NP_612189.2 | 2, 3 | 544 | ||
NM_148977.2 | NP_683878.1 | 2, 3 | 1322 | |||
NM_148978.2 | NP_683879.1 | 2, 3 | 544 | |||
XM_017016333.1 | XP_016871822.1 | 3, 4 | 1290 | |||
XM_017016334.1 | XP_016871823.1 | 3, 4 | 1252 | |||
XM_017016335.1 | XP_016871824.1 | 3, 4 | 2080 | |||
XM_017016336.1 | XP_016871825.1 | 2, 3 | 469 | |||
XM_017016337.1 | XP_016871826.1 | 3, 4 | 1986 | |||
GenBank mRNA | AF355198.1 | - | 2, 3 | 528 | ||
AK125428.1 | - | 3, 4 | 901 | |||
AK127777.1 | - | 1, 2 | 243 | |||
AY027661.1 | - | 2, 3 | 1322 | |||
AY027662.1 | - | 2, 3 | 544 | |||
AY027663.1 | - | 3, 4 | 638 |