Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005030.5 | NP_005021.2 | 9, 10 | 1777 | ||
GenBank mRNA | AB209179.1 | - | 8, 9 | 3636 | ||
AK303263.1 | - | 10, 11 | 1393 | |||
AK313227.1 | - | 9, 10 | 1701 | |||
BC002369.2 | - | 9, 10 | 1669 | 2822226 | ||
BC003002.1 | - | 9, 10 | 1647 | 2823623 | ||
BC014846.1 | - | 9, 10 | 1622 | 3854860 | ||
L19559.1 | - | 9, 10 | 1679 | |||
U01038.1 | - | 9, 10 | 1679 | |||
X73458.1 | - | 9, 10 | 1693 | |||
X75932.1 | - | 9, 10 | 1679 |