Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002401.3 | NP_002392.2 | 5, 6 | 696 | ||
NM_203351.1 | NP_976226.1 | 6, 7 | 789 | |||
XM_005257376.3 | XP_005257433.1 | 6, 7 | 1021 | |||
XM_005257377.3 | XP_005257434.1 | 5, 6 | 420 | |||
XM_005257378.2 | XP_005257435.1 | 6, 7 | 1021 | |||
GenBank mRNA | AK225784.1 | - | 6, 7 | 805 | ||
AK315305.1 | - | 6, 7 | 475 | |||
AL834303.1 | - | 6, 7 | 788 | |||
AX775739.1 | - | 7, 8 | 536 | |||
AX775741.1 | - | 7, 8 | 536 | |||
BC008336.1 | - | 4, 5 | 281 | |||
BC090859.1 | - | 5, 6 | 697 | 6183752 | ||
BC093672.1 | - | 5, 6 | 427 | 7939517 | ||
BC093674.1 | - | 5, 6 | 427 | 7939519 | ||
BC143224.1 | - | 5, 6 | 427 | |||
BC143235.1 | - | 5, 6 | 424 | |||
DQ454151.1 | - | 4, 5 | 377 | |||
U78876.1 | - | 5, 6 | 460 |