Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102406.1 | NP_001095876.1 | 4 | 391 | ||
NM_025144.3 | NP_079420.3 | 4 | 443 | |||
XM_005263245.4 | XP_005263302.1 | 4 | 443 | |||
XM_005263246.4 | XP_005263303.1 | 4 | 366 | |||
XM_006714326.3 | XP_006714389.1 | 4 | 443 | |||
XM_017008633.1 | XP_016864122.1 | 4 | 443 | |||
XM_017008634.1 | XP_016864123.1 | 3 | 411 | |||
XM_017008635.1 | XP_016864124.1 | 4 | 366 | |||
XM_017008636.1 | XP_016864125.1 | 5 | 519 | |||
XM_017008637.1 | XP_016864126.1 | 4 | 394 | |||
XM_017008638.1 | XP_016864127.1 | 4 | 532 | |||
XM_017008639.1 | XP_016864128.1 | 5 | 467 | |||
XM_017008640.1 | XP_016864129.1 | 4 | 342 | |||
XM_017008641.1 | XP_016864130.1 | 4 | 480 | |||
XM_017008642.1 | XP_016864131.1 | 4 | 443 | |||
XM_017008643.1 | XP_016864132.1 | 4 | 443 | |||
XM_017008645.1 | XP_016864134.1 | 4 | 443 | |||
XM_017008646.1 | XP_016864135.1 | 4 | 443 | |||
XM_017008650.1 | XP_016864139.1 | 4 | 333 | |||
XM_017008651.1 | XP_016864140.1 | 4 | 396 | |||
GenBank mRNA | AK026323.1 | - | 4 | 343 | ||
AK097246.1 | - | 4 | 404 | |||
AK303489.1 | - | 4 | 390 | |||
AY044164.1 | - | 2 | 246 | |||
BX648798.1 | - | 4 | 334 |