Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002401.3 | NP_002392.2 | 12 | 1451 | ||
NM_203351.1 | NP_976226.1 | 13 | 1544 | |||
XM_005257376.3 | XP_005257433.1 | 13 | 1764 | |||
XM_005257377.3 | XP_005257434.1 | 12 | 1163 | |||
XM_005257378.2 | XP_005257435.1 | 13 | 1776 | |||
GenBank mRNA | AK225784.1 | - | 13 | 1548 | ||
AK315305.1 | - | 13 | 1230 | |||
AL834303.1 | - | 13 | 1543 | |||
AX775739.1 | - | 14 | 1291 | |||
AX775741.1 | - | 14 | 1291 | |||
BC008336.1 | - | 11 | 1036 | |||
BC090859.1 | - | 12 | 1452 | 6183752 | ||
BC093672.1 | - | 12 | 1182 | 7939517 | ||
BC093674.1 | - | 12 | 1182 | 7939519 | ||
BC143224.1 | - | 12 | 1170 | |||
BC143235.1 | - | 12 | 1179 | |||
DQ454151.1 | - | 11 | 1120 | |||
U78876.1 | - | 12 | 1215 |