Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178170.2 | NP_835464.1 | 11 | 1493 | ||
XM_011524638.2 | XP_011522940.1 | 12 | 1736 | |||
XM_011524640.2 | XP_011522942.1 | 12 | 1697 | |||
XM_017024499.1 | XP_016879988.1 | 11 | 1542 | |||
XM_017024500.1 | XP_016879989.1 | 11 | 1578 | |||
GenBank mRNA | AL833909.1 | - | 2 | 163 | ||
AY242354.1 | - | 11 | 1493 | |||
AY267371.1 | - | 11 | 1493 | |||
BC112240.1 | - | 11 | 1527 | 8327708 | ||
BC113705.1 | - | 11 | 1527 | 8322757 |