Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031741.3 | NP_001026911.1 | 15 | 1629 | ||
NM_001304384.1 | NP_001291313.1 | 16 | 1659 | |||
NM_152534.4 | NP_689747.3 | 38 | 3759 | |||
XM_006712997.3 | XP_006713060.1 | 37 | 3690 | |||
XM_006712998.2 | XP_006713061.1 | 38 | 3720 | |||
XM_006712999.3 | XP_006713062.1 | 38 | 3720 | |||
XM_006713001.3 | XP_006713064.1 | 36 | 3549 | |||
XM_017005761.1 | XP_016861250.1 | 37 | 3672 | |||
XM_017005762.1 | XP_016861251.1 | 39 | 3836 | |||
XM_017005763.1 | XP_016861252.1 | 38 | 3817 | |||
XM_017005764.1 | XP_016861253.1 | 37 | 3649 | |||
XM_017005765.1 | XP_016861254.1 | 37 | 3621 | |||
XM_017005766.1 | XP_016861255.1 | 36 | 3642 | |||
XM_017005767.1 | XP_016861256.1 | 36 | 3585 | |||
XM_017005768.1 | XP_016861257.1 | 37 | 3582 | |||
XM_017005769.1 | XP_016861258.1 | 37 | 3579 | |||
XM_017005770.1 | XP_016861259.1 | 36 | 3531 | |||
XM_017005771.1 | XP_016861260.1 | 35 | 3501 | |||
XM_017005772.1 | XP_016861261.1 | 35 | 3444 | |||
GenBank mRNA | AK057247.1 | - | 16 | 1611 | ||
BC045758.1 | - | 15 | 1581 | 4821808 | ||
DQ104438.1 | - | 13 | 1211 |