Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYLK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182493.2 | NP_872299.2 | 1, 2 | 598 | ||
GenBank mRNA | AB451455.1 | - | 1, 2 | 482 | ||
AJ247087.1 | - | 1, 2 | 598 | |||
BC109097.1 | - | 1, 2 | 495 | 40034724 |