Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014238.1 | NP_055053.1 | 17 | 2174 | ||
XM_006722154.3 | XP_006722217.1 | 14 | 2218 | |||
XM_011525429.2 | XP_011523731.1 | 16 | 2326 | |||
XM_011525430.2 | XP_011523732.1 | 16 | 2328 | |||
XM_011525431.2 | XP_011523733.1 | 16 | 2328 | |||
XM_017025267.1 | XP_016880756.1 | 15 | 2284 | |||
XM_017025268.1 | XP_016880757.1 | 15 | 2260 | |||
XM_017025269.1 | XP_016880758.1 | 15 | 2260 | |||
XM_017025270.1 | XP_016880759.1 | 15 | 2061 | |||
XM_017025271.1 | XP_016880760.1 | 17 | 3033 | |||
XM_017025272.1 | XP_016880761.1 | 17 | 3016 | |||
XM_017025273.1 | XP_016880762.1 | 16 | 2839 | |||
XM_017025274.1 | XP_016880763.1 | 16 | 2042 | |||
XM_017025275.1 | XP_016880764.1 | 16 | 2029 | |||
XM_017025276.1 | XP_016880765.1 | 17 | 2981 | |||
XM_017025277.1 | XP_016880766.1 | 12 | 1724 | |||
XM_017025278.1 | XP_016880767.1 | 12 | 1734 | |||
GenBank mRNA | U43586.1 | - | 12 | 1348 |