Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000858.5 | NP_000849.1 | 4, 5 | 455 | ||
NM_001159390.1 | NP_001152862.1 | 3, 4 | 290 | |||
NM_001159391.1 | NP_001152863.1 | 3, 4 | 204 | |||
NM_001242839.1 | NP_001229768.1 | 3, 4 | 230 | |||
NM_001242840.1 | NP_001229769.1 | 3, 4 | 290 | |||
XM_005273103.3 | XP_005273160.1 | 5, 6 | 626 | |||
XM_005273104.4 | XP_005273161.1 | 5, 6 | 624 | |||
XM_005273105.3 | XP_005273162.1 | 4, 5 | 483 | |||
XM_005273106.2 | XP_005273163.1 | 3, 4 | 671 | |||
XM_011544174.2 | XP_011542476.1 | 3, 4 | 3054 | |||
GenBank mRNA | AK303845.1 | - | 3, 4 | 243 | ||
BC006249.2 | - | 4, 5 | 393 | 3946293 | ||
BC007369.2 | - | 2, 3 | 107 | |||
BC009914.2 | - | 4, 5 | 367 | 2821563 | ||
BC091480.1 | - | 3, 4 | 210 | 30340102 | ||
BE780558.1 | - | 3, 4 | 177 | |||
BI771824.1 | - | 3, 4 | 230 | |||
BU162698.1 | - | 271 | ||||
BU194785.1 | - | 256 | ||||
BU931096.1 | - | 247 | ||||
CR456836.1 | - | 2, 3 | 152 | |||
CR542006.1 | - | 2, 3 | 152 | |||
L76200.1 | - | 2, 3 | 152 | |||
U66895.1 | - | 2, 3 | 158 |