Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000601.5 | NP_000592.3 | 8, 9 | 1276 | ||
NM_001010932.2 | NP_001010932.1 | 8, 9 | 1261 | |||
XM_006715956.2 | XP_006716019.1 | 9, 10 | 1239 | |||
XM_011516115.2 | XP_011514417.1 | 9, 10 | 1224 | |||
GenBank mRNA | AK291674.1 | - | 8, 9 | 1211 | ||
AK313694.1 | - | 8, 9 | 1111 | |||
BC130284.1 | - | 8, 9 | 1124 | 40146315 | ||
BC130286.1 | - | 8, 9 | 1124 | 40146317 | ||
M29145.1 | - | 8, 9 | 1150 | |||
M55379.1 | - | 8, 9 | 1034 | |||
M60718.1 | - | 8, 9 | 1121 | |||
M73239.1 | - | 8, 9 | 1114 | |||
M73240.1 | - | 8, 9 | 1099 | |||
X16323.1 | - | 8, 9 | 1183 |