Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000930.4 | NP_000921.1 | 13 | 1660 | ||
NM_001319189.1 | NP_001306118.1 | 11 | 1393 | |||
NM_033011.3 | NP_127509.1 | 12 | 1522 | |||
GenBank mRNA | A03776.1 | - | 12 | 1437 | ||
A07197.1 | - | 12 | 1427 | |||
AF260825.1 | - | 7 | 902 | |||
AK289387.1 | - | 13 | 1612 | |||
AK290575.1 | - | 13 | 1612 | |||
AK297738.1 | - | 11 | 1234 | |||
AK297937.1 | - | 10 | 1123 | |||
AK299205.1 | - | 13 | 1502 | |||
AK300984.1 | - | 13 | 1312 | |||
AK313342.1 | - | 12 | 1363 | |||
AY221101.1 | - | 12 | 1403 | |||
BC002795.2 | - | 12 | 1348 | 3618149 | ||
BC007231.1 | - | 13 | 1484 | 3161005 | ||
BC013968.2 | - | 13 | 1486 | 3163742 | ||
BC018636.2 | - | 13 | 1486 | 3637273 | ||
BC095403.1 | - | 13 | 1501 | 30528441 | ||
BT007060.1 | - | 12 | 1403 | |||
D01096.1 | - | 11 | 1315 | |||
FM956488.1 | - | 12 | 1414 | |||
M15518.1 | - | 13 | 1479 | |||
M18182.1 | - | 12 | 1416 | |||
X02901.1 | - | 12 | 1420 | |||
X07393.1 | - | 13 | 1456 | |||
X13097.1 | - | 10 | 997 |