Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001935.3 | NP_001926.2 | 22 | 2466 | ||
XM_005246371.2 | XP_005246428.1 | 22 | 2043 | |||
GenBank mRNA | AB451339.1 | - | 21 | 1905 | ||
AB451488.1 | - | 21 | 1905 | |||
AK314798.1 | - | 23 | 2278 | |||
BC013329.2 | - | 21 | 1900 | |||
BC065265.1 | - | 22 | 2466 | 6156880 | ||
M74777.1 | - | 22 | 1915 | |||
M80536.1 | - | 21 | 1905 | |||
X60708.1 | - | 22 | 1980 |