Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002422.4 | NP_002413.1 | 4 | 719 | ||
GenBank mRNA | AK223283.1 | - | 4 | 637 | ||
AK223291.1 | - | 4 | 638 | |||
AK313310.1 | - | 4 | 635 | |||
BC069676.1 | - | 4 | 639 | 7262578 | ||
BC069716.1 | - | 4 | 639 | 7262590 | ||
BC074815.2 | - | 4 | 621 | 30915319 | ||
BC074869.2 | - | 4 | 621 | 30915468 | ||
BC105954.1 | - | 4 | 632 | 40033171 | ||
BC107490.1 | - | 4 | 632 | 40033168 | ||
BC107491.1 | - | 4 | 632 | 40033170 | ||
J03209.1 | - | 4 | 576 | |||
X05232.1 | - | 4 | 619 |