Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003365.2 | NP_003356.2 | 5 | 542 | ||
GenBank mRNA | AK129903.1 | - | 5 | 522 | ||
AK300694.1 | - | 5 | 477 | |||
AK302840.1 | - | 5 | 567 | |||
AK313090.1 | - | 5 | 512 | |||
BC009586.1 | - | 5 | 513 | 4096628 | ||
D26485.1 | - | 5 | 504 | |||
L16842.1 | - | 5 | 914 |