Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185099.1 | NP_001172028.1 | 3 | 344 | ||
NM_001185100.1 | NP_001172029.1 | 3 | 344 | |||
NM_001185101.1 | NP_001172030.1 | 3 | 344 | |||
NM_001278417.1 | NP_001265346.1 | 2 | 285 | |||
NM_001771.3 | NP_001762.2 | 3 | 344 | |||
GenBank mRNA | AK225625.1 | - | 3 | 326 | ||
AK225694.1 | - | 3 | 326 | |||
AK301177.1 | - | 3 | 320 | |||
BC109306.1 | - | 2 | 239 | 40035425 | ||
CF138699.1 | - | 3 | 323 | |||
DA931592.1 | - | 3 | 337 | |||
DA989528.1 | - | 2 | 288 | |||
X52785.1 | - | 2 | 294 | |||
X59350.1 | - | 3 | 316 |