Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPEPPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006310.3 | NP_006301.3 | 21 | 2652 | ||
XM_011525496.1 | XP_011523798.1 | 22 | 2454 | |||
XM_017025372.1 | XP_016880861.1 | 21 | 2614 | |||
XM_017025373.1 | XP_016880862.1 | 22 | 2454 | |||
GenBank mRNA | AJ132583.1 | - | 21 | 2382 | ||
AK096491.1 | - | 6 | 1110 | |||
AK096709.1 | - | 23 | 2439 | |||
AK293450.1 | - | 15 | 1584 | |||
AK293995.1 | - | 14 | 1566 | |||
AK296887.1 | - | 20 | 2295 | |||
AK297107.1 | - | 20 | 2156 | |||
AK303037.1 | - | 22 | 2447 | |||
BC065294.1 | - | 21 | 2341 | 6059589 |