Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080722.4 | NP_542453.2 | 20, 21 | 3372 | ||
NM_139155.3 | NP_631894.2 | 20, 21 | 3381 | |||
XM_011539300.2 | XP_011537602.1 | 17, 18 | 2855 | |||
XM_011539301.2 | XP_011537603.1 | 17, 18 | 2378 | |||
XM_011539302.2 | XP_011537604.1 | 17, 18 | 2255 | |||
XM_011539309.1 | XP_011537611.1 | 13, 14 | 1965 | |||
GenBank mRNA | AF358666.1 | - | 20, 21 | 3075 | ||
AF366351.1 | - | 19, 20 | 3002 |