Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK292659.1 | - | 10 | 1554 | ||
AK303388.1 | - | 11 | 1645 | |||
AK303511.1 | - | 10 | 1347 | |||
BC144094.1 | - | 11 | 1604 | |||
J05556.1 | - | 10 | 1526 |