Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GZMK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002104.2 | NP_002095.1 | 5 | 902 | ||
GenBank mRNA | BC035802.1 | - | 5 | 902 | 5748080 | |
U26174.1 | - | 5 | 898 | |||
U35237.1 | - | 5 | 858 |