Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139159.4 | NP_631898.3 | 22 | 4077 | ||
XM_005259673.2 | XP_005259730.1 | 23 | 4186 | |||
XM_011528405.1 | XP_011526707.1 | 24 | 4238 | |||
XM_011528406.1 | XP_011526708.1 | 25 | 4348 | |||
XM_011528408.1 | XP_011526710.1 | 22 | 4023 | |||
XM_011528410.1 | XP_011526712.1 | 23 | 4238 | |||
XM_017027453.1 | XP_016882942.1 | 24 | 4828 | |||
XM_017027454.1 | XP_016882943.1 | 25 | 4938 | |||
GenBank mRNA | AF452102.2 | - | 22 | 4097 | ||
AK075030.1 | - | 4 | 1678 | |||
AK131499.1 | - | 21 | 3590 | |||
AL834376.1 | - | 16 | 3027 | |||
BC000970.1 | - | 10 | 2460 | |||
BC017008.1 | - | 1 | 370 | |||
BC037948.2 | - | 22 | 4014 | 4577856 | ||
CR627380.1 | - | 14 | 2852 |