Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000083.2 | NP_000074.2 | 7, 8 | 948 | ||
NR_046453.1 | 7, 8 | 951 | ||||
XM_017011739.1 | XP_016867228.1 | 4, 5 | 649 | |||
XM_017011740.1 | XP_016867229.1 | 4, 5 | 649 | |||
GenBank mRNA | AY103154.1 | - | 6, 7 | 452 | ||
AY103155.1 | - | 5, 6 | 374 | |||
AY103156.1 | - | 2, 3 | 162 | |||
BC112156.1 | - | 7, 8 | 913 | 8327624 | ||
BC113495.1 | - | 7, 8 | 913 | 8322547 | ||
BC143796.1 | - | 7, 8 | 916 | |||
M97820.1 | - | 4, 5 | 352 | |||
Z25884.1 | - | 7, 8 | 948 |