Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000087.3 | NP_000078.2 | 11 | 1386 | ||
NM_001142564.1 | NP_001136036.1 | 10 | 1468 | |||
XM_005248049.4 | XP_005248106.2 | 9 | 1358 | |||
XM_011513623.2 | XP_011511925.1 | 10 | 1449 | |||
XM_017007712.1 | XP_016863201.1 | 11 | 1441 | |||
GenBank mRNA | AK292021.1 | - | 11 | 1386 | ||
M84741.1 | - | 9 | 1144 | |||
S42457.1 | - | 10 | 1483 |