Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CACNA1C SIRNA Assays ›
Gene Symbol:
CACNA1C
Gene Name:
calcium voltage-gated channel subunit alpha1 C
Gene Aliases:
CACH2, CACN2, CACNL1A1, CCHL1A1, CaV1.2, LQT8, TS
Chromosome Location:
Chr.12: 1969677 - 2697949 on Build GRCh38
UniGene ID:
Hs.118262
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
104331
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CACNA1C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000719.6
NP_000710.5
47
7541
NM_001129827.1
NP_001123299.1
49
7685
NM_001129829.1
NP_001123301.1
47
7664
NM_001129830.2
NP_001123302.2
48
7611
NM_001129831.1
NP_001123303.1
48
7625
NM_001129832.1
NP_001123304.1
48
7601
NM_001129833.1
NP_001123305.1
47
7598
NM_001129834.1
NP_001123306.1
47
7598
NM_001129835.1
NP_001123307.1
47
7598
NM_001129836.1
NP_001123308.1
47
7592
NM_001129837.1
NP_001123309.1
46
7565
NM_001129838.1
NP_001123310.1
46
7565
NM_001129839.1
NP_001123311.1
46
7559
NM_001129840.1
NP_001123312.1
47
7541
NM_001129841.1
NP_001123313.1
47
7541
NM_001129842.1
NP_001123314.1
47
7541
NM_001129843.1
NP_001123315.1
47
7541
NM_001129844.1
NP_001123316.1
47
7532
NM_001129846.1
NP_001123318.1
46
7508
NM_001167623.1
NP_001161095.1
47
7541
NM_001167624.2
NP_001161096.2
48
7611
NM_001167625.1
NP_001161097.1
47
7721
NM_199460.3
NP_955630.3
50
7755
XM_006719017.2
XP_006719080.1
47
7595
XM_011521020.2
XP_011519322.1
48
7670
XM_011521023.2
XP_011519325.1
48
7855
XM_017019926.1
XP_016875415.1
53
9436
XM_017019927.1
XP_016875416.1
52
9331
XM_017019928.1
XP_016875417.1
51
9118
XM_017019929.1
XP_016875418.1
51
9061
XM_017019932.1
XP_016875421.1
48
8962
XM_017019935.1
XP_016875424.1
49
8857
XM_017019946.1
XP_016875435.1
48
8782
XM_017019953.1
XP_016875442.1
47
7595
XM_017019954.1
XP_016875443.1
47
7595
XM_017019955.1
XP_016875444.1
49
8857
GenBank mRNA
AF070589.1
-
1
942
AF465484.1
-
47
7507
L29529.1
-
46
7348
L29534.1
-
47
7267
L29536.1
-
46
7162
Back To Top
More Information
Panther Classification:
Molecular Function -
voltage-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
immune system development
positive regulation of cytosolic calcium ion concentration
heart development
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
embryonic forelimb morphogenesis
calcium-mediated signaling using extracellular calcium source
camera-type eye development
regulation of insulin secretion
calcium ion transport into cytosol
cardiac conduction
calcium ion transmembrane transport via high voltage-gated calcium channel
calcium ion transmembrane transport
cardiac muscle cell action potential involved in contraction
membrane depolarization during cardiac muscle cell action potential
membrane depolarization during AV node cell action potential
cell communication by electrical coupling involved in cardiac conduction
regulation of heart rate by cardiac conduction
regulation of ventricular cardiac muscle cell action potential
membrane depolarization during atrial cardiac muscle cell action potential
voltage-gated calcium channel activity
protein binding
calmodulin binding
high voltage-gated calcium channel activity
metal ion binding
alpha-actinin binding
voltage-gated calcium channel activity involved in cardiac muscle cell action potential
voltage-gated calcium channel activity involved in AV node cell action potential
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0